Assay Design



Gene Transcripts

Gene Symbol
Pla2g4d
Entrez Gene ID
691905
Gene Name
phospholipase A2, group IVD (cytosolic)
Gene Aliases
-
Chromosome Location
Chr.3: 112040827 - 112061847 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001080051.5 XP_001080051.2 18 - 19 2118 83

View Full Product Details