Assay Design
Gene Transcripts
- Gene Symbol
- Fbln7
- Entrez Gene ID
-
296145
- Gene Name
- fibulin 7
- Gene Aliases
-
RGD1310131
- Chromosome Location
-
Chr.3: 121407992 - 121441660 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_215832.7 |
XP_215832.5 |
3 - 4 |
561 |
58 |
View Full Product Details