Assay Design
Gene Transcripts
- Gene Symbol
- Oxt
- Entrez Gene ID
-
25504
- Gene Name
- oxytocin/neurophysin 1 prepropeptide
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 123106694 - 123107534 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_012996.3 |
NP_037128.1 |
1 - 2 |
153 |
65 |
GenBank mRNA |
M25649.1 |
- |
1 - 2 |
153 |
65 |
View Full Product Details