Assay Design



Gene Transcripts

Gene Symbol
Nusap1
Entrez Gene ID
311336
Gene Name
nucleolar and spindle associated protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 111422267 - 111452600 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107762.1 NP_001101232.1 2 - 3 435 89
XM_006234768.2 XP_006234830.1 2 - 3 266 89
XM_006234769.2 XP_006234831.1 2 - 3 266 89
XM_008762101.1 XP_008760323.1 2 - 3 266 89
XM_008762102.1 XP_008760324.1 2 - 3 266 89
GenBank mRNA BC161947.1 - 2 - 3 211 89

View Full Product Details