Assay Design
Gene Transcripts
- Gene Symbol
- Trim62
- Entrez Gene ID
-
313045
- Gene Name
- tripartite motif-containing 62
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 147069355 - 147096960 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_232757.8 |
XP_232757.5 |
4 - 5 |
1144 |
109 |
View Full Product Details