Assay Design



Gene Transcripts

Gene Symbol
Pla2g2f
Entrez Gene ID
690388
Gene Name
phospholipase A2, group IIF
Gene Aliases
-
Chromosome Location
Chr.5: 157192047 - 157200167 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109587.1 NP_001103057.1 4 - 5 549 71
XM_008764289.1 XP_008762511.1 5 - 6 974 71

View Full Product Details