Assay Design



Gene Transcripts

Gene Symbol
Acot7
Entrez Gene ID
26759
Gene Name
acyl-CoA thioesterase 7
Gene Aliases
Bach
Chromosome Location
Chr.5: 169357582 - 169450215 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146061.1 NP_001139533.1 1 - 2 234 71
GenBank mRNA D88891.1 - 1 - 2 234 71

View Full Product Details