Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100709.1 | NP_001094179.1 | 4 - 5 | 381 | 85 |
XM_006238932.2 | XP_006238994.1 | 5 - 6 | 548 | 85 | |
XM_006238933.2 | XP_006238995.1 | 5 - 6 | 510 | 85 | |
XM_008764140.1 | XP_008762362.1 | 5 - 6 | 566 | 85 | |
GenBank mRNA | BC160881.1 | - | 4 - 5 | 381 | 85 |