Assay Design



Gene Transcripts

Gene Symbol
Oscp1
Entrez Gene ID
362595
Gene Name
organic solute carrier partner 1
Gene Aliases
RGD1306596
Chromosome Location
Chr.5: 144067466 - 144098073 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029923.1 NP_001025094.1 2 - 3 316 66
GenBank mRNA AB299031.1 - 2 - 3 219 66
BC098048.1 - 2 - 3 316 66

View Full Product Details