Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017271.2 | NP_058967.1 | 4 - 5 | 513 | 69 |
GenBank mRNA | BC065581.1 | - | 4 - 5 | 499 | 69 |
X82445.1 | - | 4 - 5 | 513 | 69 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008773882.1 | XP_008772104.1 | 3 - 4 | 318 | 69 |