Assay Design



Gene Transcripts

Gene Symbol
Dpf3
Entrez Gene ID
299186
Gene Name
D4, zinc and double PHD fingers, family 3
Gene Aliases
-
Chromosome Location
Chr.6: 106660569 - 106971299 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191818.1 NP_001178747.1 5 - 6 549 85
XM_008764746.1 XP_008762968.1 6 - 7 769 85
XM_008764747.1 XP_008762969.1 7 - 8 750 85
XM_008764748.1 XP_008762970.1 6 - 7 768 85
XM_008764749.1 XP_008762971.1 5 - 6 598 85
XM_008764750.1 XP_008762972.1 6 - 7 768 85
XM_008764751.1 XP_008762973.1 5 - 6 1015 85

View Full Product Details