Assay Design



Gene Transcripts

Gene Symbol
Snw1
Entrez Gene ID
500695
Gene Name
SNW domain containing 1
Gene Aliases
RGD1561926
Chromosome Location
Chr.6: 111483698 - 111507782 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109279.1 NP_001102749.1 1 - 2 70 78

View Full Product Details