Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034929.1 | NP_001030101.1 | 6 - 7 | 1040 | 85 |
XM_006240283.2 | XP_006240345.1 | 6 - 7 | 1344 | 85 | |
XM_008764784.1 | XP_008763006.1 | 5 - 6 | 1473 | 85 | |
GenBank mRNA | BC105849.1 | - | 6 - 7 | 1040 | 85 |