Assay Design



Gene Transcripts

Gene Symbol
Smek1
Entrez Gene ID
314388
Gene Name
SMEK homolog 1, suppressor of mek1 (Dictyostelium)
Gene Aliases
RGD1309059
Chromosome Location
Chr.6: 125040803 - 125083496 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108050.2 NP_001101520.2 7 - 8 1691 93
GenBank mRNA FQ087395.1 - 4 - 5 669 93

View Full Product Details