Assay Design
Gene Transcripts
- Gene Symbol
- Padi3
- Entrez Gene ID
-
29520
- Gene Name
- peptidyl arginine deiminase, type III
- Gene Aliases
-
Pdi3
- Chromosome Location
-
Chr.5: 159309000 - 159336429 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_017230.2 |
NP_058926.1 |
2 - 3 |
313 |
65 |
GenBank mRNA |
D88034.1 |
- |
2 - 3 |
314 |
65 |
View Full Product Details