Assay Design



Gene Transcripts

Gene Symbol
Srsf4
Entrez Gene ID
362612
Gene Name
serine/arginine-rich splicing factor 4
Gene Aliases
RGD1561347, Sfrs4
Chromosome Location
Chr.5: 150032999 - 150060769 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108685.2 NP_001102155.1 2 - 3 364 58

View Full Product Details