Assay Design



Gene Transcripts

Gene Symbol
Stmn1
Entrez Gene ID
29332
Gene Name
stathmin 1
Gene Aliases
Lap18, OP18, PP17, PP19, PR22, SMN, prosolin
Chromosome Location
Chr.5: 152681026 - 152686806 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006239123.2 XP_006239185.1 1 - 2 105 100

View Full Product Details