Assay Design



Gene Transcripts

Gene Symbol
Atad3a
Entrez Gene ID
298682
Gene Name
ATPase family, AAA domain containing 3A
Gene Aliases
Atad3
Chromosome Location
Chr.5: 173189590 - 173209768 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034922.1 NP_001030094.1 13 - 14 1379 86
GenBank mRNA BC105762.1 - 13 - 14 1379 86

View Full Product Details