Assay Design



Gene Transcripts

Gene Symbol
Fndc5
Entrez Gene ID
260327
Gene Name
fibronectin type III domain containing 5
Gene Aliases
-
Chromosome Location
Chr.5: 147323240 - 147330266 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270981.1 NP_001257910.1 3 - 4 416 62
GenBank mRNA AF529211.1 - 2 - 3 322 62

View Full Product Details