Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025651.1 | NP_001020822.1 | 13 - 14 | 1445 | 62 |
XM_006239013.2 | XP_006239075.1 | 13 - 14 | 2283 | 62 | |
GenBank mRNA | BC098679.1 | - | 13 - 14 | 1445 | 62 |
FQ227771.1 | - | 4 - 5 | 421 | 62 |