Assay Design



Gene Transcripts

Gene Symbol
Sytl1
Entrez Gene ID
297872
Gene Name
synaptotagmin-like 1
Gene Aliases
Slp-1
Chromosome Location
Chr.5: 151448639 - 151459875 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025651.1 NP_001020822.1 7 - 8 725 101
XM_006239013.2 XP_006239075.1 7 - 8 1563 101
GenBank mRNA BC098679.1 - 7 - 8 725 101

View Full Product Details