Assay Design



Gene Transcripts

Gene Symbol
Padi4
Entrez Gene ID
29512
Gene Name
peptidyl arginine deiminase, type IV
Gene Aliases
Pdi4
Chromosome Location
Chr.5: 159260569 - 159293694 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017227.1 NP_058923.1 11 - 12 1322 133
XM_006239145.2 XP_006239207.1 11 - 12 1343 133
GenBank mRNA AB008803.1 - 11 - 12 1322 133
AB010999.1 - 11 - 12 1322 133

View Full Product Details