Assay Design
Gene Transcripts
- Gene Symbol
- Tnfrsf4
- Entrez Gene ID
-
25572
- Gene Name
- tumor necrosis factor receptor superfamily, member 4
- Gene Aliases
-
Ox40, Txgp1
- Chromosome Location
-
Chr.5: 173447784 - 173450474 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_013049.1 |
NP_037181.1 |
5 - 6 |
640 |
88 |
GenBank mRNA |
X17037.1 |
- |
5 - 6 |
640 |
88 |
View Full Product Details