Assay Design



Gene Transcripts

Gene Symbol
Ctrc
Entrez Gene ID
362653
Gene Name
chymotrypsin C (caldecrin)
Gene Aliases
-
Chromosome Location
Chr.5: 160394677 - 160403373 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077649.2 NP_001071117.1 5 - 6 501 86
GenBank mRNA S80379.1 - 5 - 6 502 86

View Full Product Details