Assay Design



Gene Transcripts

Gene Symbol
Mfap2
Entrez Gene ID
313662
Gene Name
microfibrillar-associated protein 2
Gene Aliases
-
Chromosome Location
Chr.5: 159534202 - 159539750 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107989.1 NP_001101459.1 1 - 2 45 81

View Full Product Details