Assay Design



Gene Transcripts

Gene Symbol
Lsm10
Entrez Gene ID
366468
Gene Name
LSM10, U7 small nuclear RNA associated
Gene Aliases
-
Chromosome Location
Chr.5: 144106556 - 144108703 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238895.2 XP_006238957.1 1 - 2 117 82
GenBank mRNA BC166902.1 - 1 - 2 52 82

View Full Product Details