Assay Design



Gene Transcripts

Gene Symbol
Lsm10
Entrez Gene ID
366468
Gene Name
LSM10, U7 small nuclear RNA associated
Gene Aliases
-
Chromosome Location
Chr.5: 144106556 - 144108703 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108976.1 NP_001102446.1 1 - 2 87 83

View Full Product Details