Assay Design



Gene Transcripts

Gene Symbol
Sh2d5
Entrez Gene ID
366489
Gene Name
SH2 domain containing 5
Gene Aliases
RGD1305952
Chromosome Location
Chr.5: 156615889 - 156624962 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006239282.2 XP_006239344.1 3 - 4 616 97
XM_006239283.2 XP_006239345.1 3 - 4 274 97
XM_006239286.2 XP_006239348.1 2 - 3 117 97
XM_008764300.1 XP_008762522.1 4 - 5 604 97
XM_008764301.1 XP_008762523.1 4 - 5 607 97
XM_008764302.1 XP_008762524.1 2 - 3 104 97

View Full Product Details