Assay Design
Gene Transcripts
- Gene Symbol
- Edn2
- Entrez Gene ID
-
24324
- Gene Name
- endothelin 2
- Gene Aliases
-
Et2
- Chromosome Location
-
Chr.5: 139100898 - 139105792 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_012549.2 |
NP_036681.1 |
2 - 3 |
215 |
65 |
View Full Product Details