Assay Design
Gene Transcripts
- Gene Symbol
- Nbl1
- Entrez Gene ID
-
50594
- Gene Name
- neuroblastoma 1, DAN family BMP antagonist
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 157524569 - 157535701 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC061833.1 |
- |
1 - 2 |
|
67 |
View Full Product Details