Assay Design



Gene Transcripts

Gene Symbol
Gng4
Entrez Gene ID
114118
Gene Name
guanine nucleotide binding protein (G protein), gamma 4
Gene Aliases
-
Chromosome Location
Chr.17: 90267024 - 90316292 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751737.3 XP_003751785.1 3 - 4 573 80
XM_008771924.1 XP_008770146.1 3 - 4 505 80

View Full Product Details