Assay Design
Gene Transcripts
- Gene Symbol
- Nid1
- Entrez Gene ID
-
25494
- Gene Name
- nidogen 1
- Gene Aliases
-
Nid
- Chromosome Location
-
Chr.17: 90553161 - 90627128 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_213954.8 |
XP_213954.4 |
17 - 18 |
3502 |
82 |
View Full Product Details