Assay Design



Gene Transcripts

Gene Symbol
Clca5
Entrez Gene ID
308016
Gene Name
chloride channel calcium activated 5
Gene Aliases
Clca2
Chromosome Location
Chr.2: 250954423 - 250982696 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107450.1 NP_001100920.1 8 - 9 1575 117
XM_008761497.1 XP_008759719.1 8 - 9 2727 117

View Full Product Details