Assay Design



Gene Transcripts

Gene Symbol
Mybphl
Entrez Gene ID
310782
Gene Name
myosin binding protein H-like
Gene Aliases
RGD1310511
Chromosome Location
Chr.2: 211158917 - 211173036 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014042.1 NP_001014064.1 3 - 4 551 75
XM_006233150.2 XP_006233212.1 3 - 4 917 75
GenBank mRNA BC087113.1 - 3 - 4 551 75

View Full Product Details