Assay Design



Gene Transcripts

Gene Symbol
Rtcd1
Entrez Gene ID
295395
Gene Name
RNA terminal phosphate cyclase domain 1
Gene Aliases
-
Chromosome Location
Chr.2: 219537213 - 219558489 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004227.1 NP_001004227.1 10 - 11 1020 75
XM_008761456.1 XP_008759678.1 9 - 10 976 75
GenBank mRNA BC079374.1 - 10 - 11 1020 75

View Full Product Details