Assay Design



Gene Transcripts

Gene Symbol
Slc9b2
Entrez Gene ID
679958
Gene Name
solute carrier family 9, subfamily B (NHA2, cation proton antiporter 2), member 2
Gene Aliases
Nhedc2
Chromosome Location
Chr.2: 240486054 - 240519150 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109385.2 NP_001102855.1 9 - 10 1316 81
GenBank mRNA BC166815.1 - 9 - 10 1316 81

View Full Product Details