Assay Design
Gene Transcripts
- Gene Symbol
- Negr1
- Entrez Gene ID
-
59318
- Gene Name
- neuronal growth regulator 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 262914240 - 263650441 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_021682.1 |
NP_067714.1 |
6 - 7 |
1010 |
89 |
GenBank mRNA |
AB017139.1 |
- |
6 - 7 |
1010 |
89 |
View Full Product Details