Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135879.1 | NP_001129351.1 | 1 - 2 | 156 | 126 |
XM_006233244.2 | XP_006233306.1 | 3 - 4 | 430 | 126 | |
XM_006233246.2 | XP_006233308.1 | 3 - 4 | 433 | 126 | |
XM_006233251.2 | XP_006233313.1 | 3 - 4 | 253 | 126 | |
XM_006233252.2 | XP_006233314.1 | 2 - 3 | 201 | 126 | |
XM_006233253.2 | XP_006233315.1 | 2 - 3 | 335 | 126 | |
XM_006233254.2 | XP_006233316.1 | 2 - 3 | 338 | 126 | |
XM_008761467.1 | XP_008759689.1 | 2 - 3 | 600 | 126 | |
XM_008761468.1 | XP_008759690.1 | 2 - 3 | 234 | 126 |