Assay Design



Gene Transcripts

Gene Symbol
Unc5c
Entrez Gene ID
362049
Gene Name
unc-5 homolog C (C. elegans)
Gene Aliases
-
Chromosome Location
Chr.2: 247045813 - 247397483 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199407.1 NP_955439.1 15 - 16 2781 69
XM_006233402.2 XP_006233464.1 16 - 17 2786 69
GenBank mRNA AB118026.1 - 15 - 16 2781 69

View Full Product Details