Assay Design



Gene Transcripts

Gene Symbol
Lppr5
Entrez Gene ID
310812
Gene Name
lipid phosphate phosphatase-related protein type 5
Gene Aliases
PRG-5, Pap2d, RGD1309567
Chromosome Location
Chr.2: 219895141 - 220102998 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107720.1 NP_001101190.1 1 - 2 841 108
XM_008761462.1 XP_008759684.1 1 - 2 2048 108
GenBank mRNA EU792473.1 - 1 - 2 841 108

View Full Product Details