Assay Design



Gene Transcripts

Gene Symbol
Mcoln3
Entrez Gene ID
308022
Gene Name
mucolipin 3
Gene Aliases
-
Chromosome Location
Chr.2: 251930532 - 252007722 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012059.1 NP_001012059.1 3 - 4 544 72
GenBank mRNA BC079200.1 - 3 - 4 544 72

View Full Product Details