Assay Design



Gene Transcripts

Gene Symbol
Frrs1
Entrez Gene ID
310810
Gene Name
ferric-chelate reductase 1
Gene Aliases
Sdfr2
Chromosome Location
Chr.2: 220415567 - 220469420 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008761474.1 XP_008759696.1 8 - 9 1016 106
GenBank mRNA FQ210266.1 - 8 - 9 953 106

View Full Product Details