Assay Design



Gene Transcripts

Gene Symbol
Metap1
Entrez Gene ID
295500
Gene Name
methionyl aminopeptidase 1
Gene Aliases
-
Chromosome Location
Chr.2: 243744620 - 243777673 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106476.1 NP_001099946.1 1 - 2 198 56
GenBank mRNA FQ225038.1 - 1 - 2 121 56
FQ227458.1 - 1 - 2 121 56

View Full Product Details