Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107714.1 | NP_001101184.1 | 6 - 7 | 715 | 82 |
XM_006233114.2 | XP_006233176.1 | 6 - 7 | 664 | 82 | |
XM_008761396.1 | XP_008759618.1 | 5 - 6 | 1111 | 82 | |
XM_008761397.1 | XP_008759619.1 | 5 - 6 | 1111 | 82 | |
XM_008761398.1 | XP_008759620.1 | 6 - 7 | 664 | 82 | |
XM_008761399.1 | XP_008759621.1 | 6 - 7 | 748 | 82 | |
GenBank mRNA | BC158880.1 | - | 6 - 7 | 651 | 82 |