Assay Design



Gene Transcripts

Gene Symbol
Kat2b
Entrez Gene ID
301164
Gene Name
K(lysine) acetyltransferase 2B
Gene Aliases
Pcaf
Chromosome Location
Chr.9: 4440982 - 4493791 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750617.3 XP_003750665.2 7 - 8 2123 55

View Full Product Details