Assay Design



Gene Transcripts

Gene Symbol
Dym
Entrez Gene ID
291433
Gene Name
dymeclin
Gene Aliases
RGD1309111
Chromosome Location
Chr.18: 70996075 - 71313033 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106133.2 NP_001099603.2 16 - 17 2011 87
XM_008772103.1 XP_008770325.1 15 - 16 2073 87
XM_008772104.1 XP_008770326.1 15 - 16 2073 87
GenBank mRNA BC098944.1 - 8 - 9 1021 87
BC168151.1 - 15 - 16 2002 87
CO558582.1 - 2 - 3 224 87
DV722630.1 - 5 - 6 572 87

View Full Product Details