Assay Design



Gene Transcripts

Gene Symbol
Dym
Entrez Gene ID
291433
Gene Name
dymeclin
Gene Aliases
RGD1309111
Chromosome Location
Chr.18: 70996075 - 71313033 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106133.2 NP_001099603.2 7 - 8 882 132
XM_008772103.1 XP_008770325.1 7 - 8 947 132
XM_008772104.1 XP_008770326.1 7 - 8 947 132
GenBank mRNA BC168151.1 - 7 - 8 876 132
EV765460.1 - - 368 132

View Full Product Details