Assay Design



Gene Transcripts

Gene Symbol
Sft2d1
Entrez Gene ID
292305
Gene Name
SFT2 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 52998548 - 53014444 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008302.1 NP_001008303.1 2 - 3 218 64
GenBank mRNA BC085346.1 - 2 - 3 218 64
FQ225074.1 - 2 - 3 239 64
FQ227930.1 - 2 - 3 196 64

View Full Product Details