Assay Design



Gene Transcripts

Gene Symbol
Csn1s2a
Entrez Gene ID
114595
Gene Name
casein alpha s2-like A
Gene Aliases
Csng
Chromosome Location
Chr.14: 21930858 - 21944921 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105741.1 NP_001099211.1 5 - 6 193 58

View Full Product Details