Assay Design
Gene Transcripts
- Gene Symbol
- Sult1d1
- Entrez Gene ID
-
60393
- Gene Name
- sulfotransferase family 1D, member 1
- Gene Aliases
-
ST1D1, Sult-n, Sultn
- Chromosome Location
-
Chr.14: 22107418 - 22126011 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_021769.1 |
NP_068537.1 |
5 - 6 |
621 |
159 |
GenBank mRNA |
U32372.1 |
- |
5 - 6 |
621 |
159 |
View Full Product Details